missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACVR2A Protein
A cDNA sequence encoding the ACVR2A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10955-1mg
Additional Details : Weight : 0.01000kg
Specifications
ACVR2A Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
AILGRSETQE CLFFNANWEK DRTNQTGVEP CYGDKDKRRH CFATWKNISG SIEIVKQGCW LDDINCYDRT DCVEKKDSPE VYFCCCEGNM CNEKFSYFPE MEVTQPTSNP VTPKPPLEHH HHHH | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ACVR2A protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |