missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Activin RIIB Fc Chimera (Sf 21)
Description
The Recombinant Human Activin RIIB Fc Chimera (Sf 21) is derived from Sf 21 (baculovirus). The Recombinant Human Activin RIIB Fc Chimera (Sf 21) has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 93 |
| Molecular Weight (g/mol) | 41 kDa (monomer) |
| Name | Activin RIIB Fc Chimera |
| Quantity | 100 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Activin RIIB protein, (N-terminus) Met Human Activin RIIB (Ser19-Thr 134) Accession &Num; CAA54671.1 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Gene Alias | activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ActivinRIIB, ACTRIIB, ACTR-IIB, ACVR2B, EC 2.7.11, EC 2.7.11.30, MGC116908 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?