missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Activin AC Heterodimer Protein
Description
The Recombinant Human Activin AC Heterodimer Protein is derived from CHO. The Recombinant Human Activin AC Heterodimer Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
| Molecular Weight (g/mol) | 12.9 kDa (betaA subunit), 12.5 kDa (betaC subunit) |
| Name | Activin AC |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Activin AC protein, (N-terminus) Human Activin beta A (Gly311-Ser426) Accession &Num; NP_002183 Human Activin beta C (Gly237-Ser352) Accession &Num; NP_005529 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?