missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Activin A (CHO derived) Protein

Product Code. 18746335
Click to view available options
Quantity:
1 mg
100 μg
20 μg
50 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18746335

Brand: R&D Systems™ 11348AC01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.200-1.60 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08476.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 3624
Molecular Weight (g/mol) MolecularWeight-observed: 13 kDa under reducing conditions and 24 kDa under non-reducing conditions., MolecularWeight-theroretical: 13 kDa
Quantity 1 mg
Source Chinese Hamster Ovary cell line, CHO-derived human Activin A protein Gly311-Ser426
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias activin AB alpha polypeptide, Activin beta-A chain, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FSH-releasing protein, inhibin beta A chain, inhibin beta A subunit, Inhibin, beta-1
Conjugate Unconjugated
Reconstitution Reconstitute 20 μg size at 100 μg/mL in sterile 4 mM HCl. Reconstitute all the other sizes (at 500 μg/mL in sterile 4 mM HCl.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Activin A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.