missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Active Src Protein
Description
The specific activity of Src was determined to be 100 nmol/min/mg using a synthetic peptide substrate.
Specifications
Specifications
| Accession Number | NM_005417 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied in 50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol. |
| Gene ID (Entrez) | 6714 |
| Molecular Weight (g/mol) | M.W. (Observed): 83 kDa |
| Quantity | 10 μg |
| Source | E. coli-derived human Src protein Accession # NM_005417 |
| Storage Requirements | This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.) |
| Gene Symbol | SRC |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?