missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Active FAK Protein

Product Code. 18157180
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18157180

Brand: R&D Systems™ 4467KS010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The specific activity of FAK was determined to be 51 nmol/min/mg as per Activity Assay Protocol, and was equivalent to 291.5 nmol/min/mg as per radiometric assay.
TRUSTED_SUSTAINABILITY

Specifications

Components Supplied in 50mM sodium phosphate (pH 7.0), 300mM NaCl, 0.25mM DTT, 150mM imidazole, 0.1mM PMSF, and 25% glycerol.
Purity >75%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
For Use With (Application) Bioactivity
Storage Buffer Supplied in 50 mM sodium phosphate (pH 7.0), 300 mM NaCl, 150 mM Imidazole, 0.1 mM PMSF, 0.25 mM DTT, and 25% Glycerol.
Content And Storage This product is stable at <= -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store (at recommended temperature. Avoid repeated freeze-thaw cycles.)
Source Spodoptera frugiperda, Sf 9 (baculovirus)-derived human FAK protein
Quantity 10 μg
Product Type Recombinant Human Focal adhesion kinase 1
Molecular Weight (g/mol) Observed MW: 35kDa
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.