missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Active Coagulation Factor XIV/ProteinC
Click to view available options
:
10ug; Carrier Free
Description
The Recombinant Human Active Coagulation Factor XIV/ProteinC is derived from CHO. The Recombinant Human Active Coagulation Factor XIV/ProteinC has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 5624 |
| Molecular Weight (g/mol) | 19 kDa (light chain), 29 kDa (heavy chain) |
| Name | Active Coagulation Factor XIV/Protein C |
| Quantity | 10μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Coagulation Factor XIV/Protein C protein Ala43-Pro461,with a C-terminal 10-His tag The isolated protein was activated by thrombin/thrombomodulin complex and further purified. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction