missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Active Caspase-2 Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Recombinant Human Active Caspase-2 Protein is derived from E. coli. The Recombinant Human Active Caspase-2 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose. |
| Gene ID (Entrez) | 835 |
| Molecular Weight (g/mol) | 12 kDa & 18 kDa |
| Name | Active Caspase-2 |
| Quantity | 10 μg |
| Source | E. coli-derived human Caspase-2 protein Gly170-Asp333 & Ala348-Tyr452 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction