missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Actin, alpha skeletal muscle Protein
A DNA sequence encoding the Homo sapiens (Human) Actin, alpha skeletal muscle, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5610-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001091.1 | |
Actin, alpha skeletal muscle Protein | |
Research Use Only | |
ACTA1 | |
Recombinant Protein | |
E. coli | |
DEDETTALVC DNGSGLVKAG FAGDDAPRAV FPSIVGRPRH QGVMVGMGQK DSYVGDEAQS KRGILTLKYP IEHGIITNWD DMEKIWHHTF YNELRVAPEE HPTLLTEAPL NPKANREKMT QIMFETFNVP AMYVAIQAVL SLYASGRTTG IVLDSGDGVT HNVPIYEGYA LPHAIMRLDL AGRDLTDYLM KILTERGYSF VTTAEREIVR DIKEKLCYVA LDFENEMATA ASSSSLEKSY ELPDGQVITI GNERFRCPET LFQPSFIGME SAGIHETTYN SIMKCDIDIR KDLYANNVMS GGTTMYPGIA DRMQKEITAL APSTMKIKII APPERKYSVW IGGSILASLS TFQQMWITKQ EYDEAGPSIV HRKCF | |
Greater than 90% as determined by SDS-PAGE. |
58 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |