missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human ACSM3 GST (N-Term) Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Gene ID (Entrez) | 6296 |
| Molecular Weight (g/mol) | 36.63 kDa |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | acyl-CoA synthetase medium-chain family member 3Protein SA homolog, acyl-coenzyme A synthetase ACSM3, mitochondrial, Butyrate--CoA ligase 3, Butyryl-coenzyme A synthetase 3, EC 6.2.1, Middle-chain acyl-CoA synthetase 3, SA, SA (rat hypertension-associated) homolog, SA hypertension-associated homolog, SA hypertension-associated homolog (rat), SAH, SAHEC 6.2.1.2 |
| Gene Symbol | ACSM3 |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Protein | ACSM3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction