missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ACP6 Protein
Description
Measured by its ability to cleave a substrate, p-Nitrophenyl phosphate (pNPP). The specific activity is >1800 pmol/min/μg, as measured under the described conditions.
Spécification
Spécification
| Accession Number | BAA91310 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 51205 |
| Molecular Weight (g/mol) | M.W. (Observed): 43-45 kDa, reducing conditions; M.W. (theoretical): 46 kDa |
| Name | ACP6 |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human ACP6 protein Glu33-Glu428, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?