missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACO1 Protein

Product Code. 15925638
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 15925638

Brand: enQuireBio™ QP109405ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the ACO1 was constructed and used to recombinantly synthesize the protein.

Specifications

Name ACO1 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMSNPFAH LAEPLDPVQP GKKFFNLNKL EDSRYGRLPF SIRVLLEAAI RNCDEFLVKK QDIENILHWN VTQHKNIEVP FKPARVILQD FTGVPAVVDF AAMRDAVKKL GGDPEKINPV CPADLVIDHS IQVDFNRRAD SLQKNQDLEF ERNRERFEFL KWGSQAFHNM RIIPPGSGII HQVNLEYLAR VVFDQDGYYY PDSLVGTDSH TTMIDGLGIL GWGVGGIEAE AVMLGQPISM VLPQVIGYRL MGKPHPLVTS TDIVLTITKH LRQVGVVGKF VEFFGPGVAQ LSIADRATIA NMCPEYGATA AFFPVDEVSI TYLVQTGRDE EKLKYIKKYL QAVGMFRDFN DPSQDPDFTQ VVELDLKTVV PCCSGPKRPQ DKVAVSDMKK DFESCLGAKQ GFKGFQVAPE HHNDHKTFIY DNTEFTLAHG SVVIAAITSC TNTSNPSVML GAGLLAKKAV DAGLNVMPYI KTSLSPGSGV VTYYLQESGV MPYLSQLGFD VVGYGCMTCI GNSGPLPEPV VEAITQGDLV AVGVLSGNRN FEGRVHPNTR ANYLASPPLV IAYAIAGTIR IDFEKEPLGV NAKGQQVFLK DIWPTRDEIQ AVERQYVIPG MFKEVYQKIE TVNESWNALA TPSDKLFFWN SKSTYIKSPP FFENLTLDLQ PPKSIVDAYV LLNLGDSVTT DHISPAGNIA RNSPAARYLT NRGLTPREFN SYGSRRGNDA VMARGTFANI RLLNRFLNKQ APQTIHLPSG EILDVFDAAE RYQQAGLPLI VLAGKEYGAG SSRDWAAKGP FLLGIKAVLA ESYERIHRSN LVGMGVIPLE YLPGENADAL GLTGQERYTI IIPENLKPQM KVQVKLDTGK TFQAVMRFDT DVELTYFLNG GILNYMIRKM AK
Buffer The ACO1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)2mM DTT, 100mM NaCl and 10% glycerol.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.