missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACO1 Protein
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
Specifications
Specifications
| Name | ACO1 Protein |
| Quantity | 1 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMSNPFAH LAEPLDPVQP GKKFFNLNKL EDSRYGRLPF SIRVLLEAAI RNCDEFLVKK QDIENILHWN VTQHKNIEVP FKPARVILQD FTGVPAVVDF AAMRDAVKKL GGDPEKINPV CPADLVIDHS IQVDFNRRAD SLQKNQDLEF ERNRERFEFL KWGSQAFHNM RIIPPGSGII HQVNLEYLAR VVFDQDGYYY PDSLVGTDSH TTMIDGLGIL GWGVGGIEAE AVMLGQPISM VLPQVIGYRL MGKPHPLVTS TDIVLTITKH LRQVGVVGKF VEFFGPGVAQ LSIADRATIA NMCPEYGATA AFFPVDEVSI TYLVQTGRDE EKLKYIKKYL QAVGMFRDFN DPSQDPDFTQ VVELDLKTVV PCCSGPKRPQ DKVAVSDMKK DFESCLGAKQ GFKGFQVAPE HHNDHKTFIY DNTEFTLAHG SVVIAAITSC TNTSNPSVML GAGLLAKKAV DAGLNVMPYI KTSLSPGSGV VTYYLQESGV MPYLSQLGFD VVGYGCMTCI GNSGPLPEPV VEAITQGDLV AVGVLSGNRN FEGRVHPNTR ANYLASPPLV IAYAIAGTIR IDFEKEPLGV NAKGQQVFLK DIWPTRDEIQ AVERQYVIPG MFKEVYQKIE TVNESWNALA TPSDKLFFWN SKSTYIKSPP FFENLTLDLQ PPKSIVDAYV LLNLGDSVTT DHISPAGNIA RNSPAARYLT NRGLTPREFN SYGSRRGNDA VMARGTFANI RLLNRFLNKQ APQTIHLPSG EILDVFDAAE RYQQAGLPLI VLAGKEYGAG SSRDWAAKGP FLLGIKAVLA ESYERIHRSN LVGMGVIPLE YLPGENADAL GLTGQERYTI IIPENLKPQM KVQVKLDTGK TFQAVMRFDT DVELTYFLNG GILNYMIRKM AK |
| Buffer | The ACO1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)2mM DTT, 100mM NaCl and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction