missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human ACE-2 Fc Chimera Protein

Product Code. 18728136
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
This item is not returnable. View return policy

Product Code. 18728136

Brand: R&D Systems™ 10544ZN020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bioactive, High Spike RBD Binding

Measured by its binding ability in a functional ELISA with Recombinant Viral SARS-CoV-2 S Protein RBD Fc Chimera (Catalog # 10499-CV ). Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH (Catalog # ES007 ). The specific activity is >550 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BYF1.2
For Use With (Application) Bioactivity, Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, ZnCl2 and Glycerol.
Gene ID (Entrez) 59272
Molecular Weight (g/mol) M.W.-observed: 124-130 kDa, under reducing conditions, M.W.-theoretical: 110 kDa
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human ACE-2 protein Human ACE-2 (Gln18-Ser740) (N-terminus) Accession # Q9BYF1.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ACE-2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.