missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACADL Protein

A cDNA sequence encoding the ACADL was constructed and used to recombinantly synthesize the protein.

£98.39 - £5741.17

Specifications

Name ACADL Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15985608
View Documents Promotion Details
enQuireBio™
QP10932-2UG
2 μg
£98.39
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15965608
View Documents Promotion Details
enQuireBio™
QP10932-10UG
10 μg
£151.37
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15975608
View Documents Promotion Details
enQuireBio™
QP10932-1MG
1 mg
£5741.17
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

ACADL Protein
His
Human
Greater than 85% as determined by SDS-PAGE.
ACADL
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGGEERLETP SAKKLTDIGI RRIFSPEHDI FRKSVRKFFQ EEVIPHHSEW EKAGEVSREV WEKAGKQGLL GVNIAEHLGG IGGDLYSAAI VWEEQAYSNC SGPGFSIHSG IVMSYITNHG SEEQIKHFIP QMTAGKCIGA IAMTEPGAGS DLQGIKTNAK KDGSDWILNG SKVFISNGSL SDVVIVVAVT NHEAPSPAHG ISLFLVENGM KGFIKGRKLH KMGLKAQDTA ELFFEDIRLP ASALLGEENK GFYYIMKELP QERLLIADVA ISASEFMFEE TRNYVKQRKA FGKTVAHLQT VQHKLAELKT HICVTRAFVD NCLQLHEAKR LDSATACMAK YWASELQNSV AYDCVQLHGG WGYMWEYPIA KAYVDARVQP IYGGTNEIMK ELIAREIVFD K
33
SDS
Product Certifications