missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACAA1 / PTHIO Protein
A DNA sequence encoding the Homo sapiens (Human) ACAA1 / PTHIO, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5602-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001123882.1 | |
ACAA1 / PTHIO Protein | |
Research Use Only | |
ACAA1 | |
Recombinant Protein | |
E. coli | |
LSGAPQASAA DVVVVHGRRT AICRAGRGGF KDTTPDELLS AVMTAVLKDV NLRPEQLGDI CVGNVLQPGA GAIMARIAQF LSDIPETVPL STVNRQCSSG LMS-QAVASI AGGIRNGSYD IGMACGITSE NVAERFGISR EKQDTFALAS QQKAARAQSK GCFQAEIVPV TTTVHDDKGT KRSITVTQDE GIRPSTTMEG LAKLKPAFKK DGSTTAGLTV SDVDIFEINE AFASQAAYCV EKLRLPPEKV NPLGGAVALG HPLGCTGARQ VITLLNELKR RGKRAYGVVS MCIGTGMGAA AVFEYPGN | |
Greater than 90% as determined by SDS-PAGE. |
30 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |