missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ABP1/AOC1 Protein
Click to view available options
:
10ug
Description
Measured by its ability to produce hydrogen peroxide during the oxidation of histamine. The specific activity is >40 pmol/min/μg, as measured under the described conditions.
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately
Specifications
Specifications
| Accession Number | CAA55046 |
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl, Glycerol |
| Molecular Weight (g/mol) | 84 kDa |
| Name | ABP1/AOC1, Carrier Free |
| Quantity | 10μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human ABP1/AOC1 protein Glu20-Val751,with a N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Gene Alias | ABP, amiloride binding protein 1 (amine oxidase (copper-containing)), Amiloride-binding protein, amiloride-sensitive amine oxidase, amiloride-sensitive amine oxidase [copper-containing], AOC1, AOC1DAOamiloride-binding protein-1, DAO1, Diamine oxidase, EC 1.4.3, EC 1.4.3.22, Histaminase, KAO, Kidney amine oxidase |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction