missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human A2M Protein

A cDNA sequence encoding the A2M was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10922-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15985578

  • £99.15 / 200µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

2
200 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Human
MGKNKLLHPS LVLLLLVLLP TDASVSGKPQ YMVLVPSLLH TETTEKGCVL LSYLNETVTV SASLESVRGN RSLFTDLEAE NDVLHCVAFA VPKSSSNEEV MFLTVQVKGP TQEFKKRTTV MVKNEDSLVF VQTDKSIYKP GQTVKFRVVS MDENFHPLNE LIPLVYIQDP KGNRIAQWQS FQLEGGLKQF SFPLSSEPFQ GSYKVVVQKK SGGRTEHPFT VEEFVLPKFE VQVTVPKIIT ILEEEMNVSV CGLYTYGKPV PGHVTVSICR KYSDASDCHG EDSQAFCEKF SGQLNSHGCF YQQVKTKVFQ LKRKEYEMKL HTEAQIQEEG TVVELTGRQS SEITRTITKL SFVKVDSHFR QGIPFFGQVR LVDGKGVPIP NKVIFIRGNE ANYYSNATTD EHGLVQFSIN TTNVMGTSLT VRVNYKDRSP CYGYQWVSEE HEEAHHTAYL VFSPSKSFVH LEPMSHELPC GHTQTVQAHY ILNGGTLLGL KKLSFYYLIM AKGGIVRTGT HGLLVKQEDM KGHFSISIPV KSDIAPVARL LIYAVLPTGD VIGDSAKYDV ENCLANKVDL SFSPSQSLPA SHAHLRVTAA PQSVCALRAV DQSVLLMKPD AELSASSVYN LLPEKDLTGF PGPLNDQDNE DCINRHNVYI NGITYTPVSS TNEKDMYSFL EDMGLKAFTN SKIRKPKMCP QLQQYEMHGP EGLRVGFYES DVMGRGHARL VHVEEPHTET VRKYFPETWI WDLVVVNSAG VAEVGVTVPD TITEWKAGAF CLSEDAGLGI SSTASLRAFQ PFFVELTMPY SVIRGEAFTL KATVLNYLPK CIRVSVQLEA SPAFLAVPVE KEQAPHCICA NGRQTVSWAV TPKSLGNVNF TVSAEALESQ ELCGTEVPSV PEHGRKDTVI KPLLVEPEGL
Greater than 96.0%.
A2M Protein
Research Use Only
A2M
Human
Untagged
Lyophilized from 0.02M TRIS, 0.15M NaCl and 0.1M Sucrose, pH 8.0.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers