missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human A2M Protein
A cDNA sequence encoding the A2M was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10922-1mg
Additional Details : Weight : 0.01000kg
Specifications
2 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
MGKNKLLHPS LVLLLLVLLP TDASVSGKPQ YMVLVPSLLH TETTEKGCVL LSYLNETVTV SASLESVRGN RSLFTDLEAE NDVLHCVAFA VPKSSSNEEV MFLTVQVKGP TQEFKKRTTV MVKNEDSLVF VQTDKSIYKP GQTVKFRVVS MDENFHPLNE LIPLVYIQDP KGNRIAQWQS FQLEGGLKQF SFPLSSEPFQ GSYKVVVQKK SGGRTEHPFT VEEFVLPKFE VQVTVPKIIT ILEEEMNVSV CGLYTYGKPV PGHVTVSICR KYSDASDCHG EDSQAFCEKF SGQLNSHGCF YQQVKTKVFQ LKRKEYEMKL HTEAQIQEEG TVVELTGRQS SEITRTITKL SFVKVDSHFR QGIPFFGQVR LVDGKGVPIP NKVIFIRGNE ANYYSNATTD EHGLVQFSIN TTNVMGTSLT VRVNYKDRSP CYGYQWVSEE HEEAHHTAYL VFSPSKSFVH LEPMSHELPC GHTQTVQAHY ILNGGTLLGL KKLSFYYLIM AKGGIVRTGT HGLLVKQEDM KGHFSISIPV KSDIAPVARL LIYAVLPTGD VIGDSAKYDV ENCLANKVDL SFSPSQSLPA SHAHLRVTAA PQSVCALRAV DQSVLLMKPD AELSASSVYN LLPEKDLTGF PGPLNDQDNE DCINRHNVYI NGITYTPVSS TNEKDMYSFL EDMGLKAFTN SKIRKPKMCP QLQQYEMHGP EGLRVGFYES DVMGRGHARL VHVEEPHTET VRKYFPETWI WDLVVVNSAG VAEVGVTVPD TITEWKAGAF CLSEDAGLGI SSTASLRAFQ PFFVELTMPY SVIRGEAFTL KATVLNYLPK CIRVSVQLEA SPAFLAVPVE KEQAPHCICA NGRQTVSWAV TPKSLGNVNF TVSAEALESQ ELCGTEVPSV PEHGRKDTVI KPLLVEPEGL | |
Greater than 96.0%. |
A2M Protein | |
Research Use Only | |
A2M | |
Human | |
Untagged | |
Lyophilized from 0.02M TRIS, 0.15M NaCl and 0.1M Sucrose, pH 8.0. |