missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human 5T4 Fc Chimera Biotinylated Protein, CF
Description
Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human 5T4 Fc Chimera (Catalog # BT4975) binds Human 5T4 Antibody (Catalog # AF4975 ) with an ED50 of 1.20-12.0 ng/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 7162 |
| Molecular Weight (g/mol) | M.W.-observed: 90-105 kDa, under reducing conditions., M.W.-theroretical: 62 kDa |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human 5T4 protein Human 5T4 (Ser31-Ser355) Accession # (N-terminus) Q13641 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, M6P1, TPBG, trophoblast glycoprotein |
| Conjugate | Biotin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?