missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 43349 Protein
A cDNA sequence encoding the 43349 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10915-20ug
Additional Details : Weight : 0.01000kg
Specifications
43349 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAATDIA RQVGEGCRTV PLAGHVGFDS LPDQLVNKSV SQGFCFNILC VGETGLGKST LMDTLFNTKF EGEPATHTQP GVQLQSNTYD LQESNVRLKL TIVSTVGFGD QINKEDSYKP IVEFIDAQFE AYLQEELKIR RVLHTYHDSR IHVCLYFIAP TGHSLKSLDL VTMKKLDSKV NIIPIIAKAD AISKSELTKF KIKITSELVS NGVQIYQFPT DDESVAEING TMNAHLPFAV IGSTEELKIG NKMMRARQYP WGTVQVENEA HCDFVKLREM LIRVNMEDLR EQTHTRHYEL YRRCKLEEMG FKDTDPDSKP FSLQETYEAK RNEFLGELQK KEEEMRQMFV QRVKEKEAEL KEAEKELHEK FDRLKKLHQD EKKKLEDKKK SLDDEVNAFK QRKTAAELPQ SQGSQAGGSQ TLKRDKEKKN NPWLCTE | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SEPT6 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 0.1M NaCl and 10% glycerol. |