missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 43349 Protein

A cDNA sequence encoding the 43349 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10915-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955558

  • £2975.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

43349 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAATDIA RQVGEGCRTV PLAGHVGFDS LPDQLVNKSV SQGFCFNILC VGETGLGKST LMDTLFNTKF EGEPATHTQP GVQLQSNTYD LQESNVRLKL TIVSTVGFGD QINKEDSYKP IVEFIDAQFE AYLQEELKIR RVLHTYHDSR IHVCLYFIAP TGHSLKSLDL VTMKKLDSKV NIIPIIAKAD AISKSELTKF KIKITSELVS NGVQIYQFPT DDESVAEING TMNAHLPFAV IGSTEELKIG NKMMRARQYP WGTVQVENEA HCDFVKLREM LIRVNMEDLR EQTHTRHYEL YRRCKLEEMG FKDTDPDSKP FSLQETYEAK RNEFLGELQK KEEEMRQMFV QRVKEKEAEL KEAEKELHEK FDRLKKLHQD EKKKLEDKKK SLDDEVNAFK QRKTAAELPQ SQGSQAGGSQ TLKRDKEKKN NPWLCTE
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The SEPT6 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers