missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 40S ribosomal protein S27 Protein

A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S27, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6626-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084640

  • £2480.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001021.1
40S ribosomal protein S27 Protein
Research Use Only
RPS27
Recombinant Protein
E. coli
PLAKDLLHPS PEEEKRKHKK KRLVQSPNSY FMDVKCPGCY KITTVFSHAQ TVVLCVGCST VLCQPTGGKA RLTEGCSFRR KQH
Greater than 80% as determined by SDS-PAGE.
6232
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers