missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 40S ribosomal protein S27 Protein

A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S27, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6626-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16014650

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001021.1
40S ribosomal protein S27 Protein
Research Use Only
RPS27
Recombinant Protein
E. coli
PLAKDLLHPS PEEEKRKHKK KRLVQSPNSY FMDVKCPGCY KITTVFSHAQ TVVLCVGCST VLCQPTGGKA RLTEGCSFRR KQH
Greater than 80% as determined by SDS-PAGE.
6232
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers