missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 40S ribosomal protein S13 Protein

A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S13, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8411-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064461

  • £384.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001008.1
40S ribosomal protein S13 Protein
Research Use Only
RPS13
Recombinant Protein
E. coli
RMHAPGKGLS QSALPYRRSV PTWLKLTSDD VKEQIYKLAK KGLTPSQIGV ILRDSHGVAQ VRFVTGNKIL RILKSKGLAP DLPEDLYHLI KKAVAVRKHL ERNRKDKDAK FRLILIESRI HRLARYYKTK RVLPPNWKYE SSTASALVA
Greater than 80% as determined by SDS-PAGE.
6207
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers