missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 4-1BBL / CD137L / TNFSF9 Protein

A DNA sequence encoding the Homo sapiens (Human) 4-1BBL / CD137L / TNFSF9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8594-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16025481

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003802.1
4-1BBL / CD137L / TNFSF9 Protein
Research Use Only
TNFSF9
Recombinant Protein
E. coli
PWAVSGARAS PGSAASPRLR EGPELSPDDP AGLLDLRQGM FAQLVAQNVL LIDGPLSWYS DPGLAGVSLT GGLSYKEDTK ELVVAKAGVY YVFFQLELRR VVAGEGSGSV SLALHLQPLR SAAGAAALAL TVDLPPASSE ARNSAFGFQG RLLHLSAGQR LGVHLHTEAR ARHAWQLTQG ATVLGLFRVT PEIPAGLPSP RSE
Greater than 80% as determined by SDS-PAGE.
8744
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers