missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 4-1BBL / CD137L / TNFSF9 Protein
A DNA sequence encoding the Homo sapiens (Human) 4-1BBL / CD137L / TNFSF9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8594-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003802.1 | |
4-1BBL / CD137L / TNFSF9 Protein | |
Research Use Only | |
TNFSF9 | |
Recombinant Protein | |
E. coli | |
PWAVSGARAS PGSAASPRLR EGPELSPDDP AGLLDLRQGM FAQLVAQNVL LIDGPLSWYS DPGLAGVSLT GGLSYKEDTK ELVVAKAGVY YVFFQLELRR VVAGEGSGSV SLALHLQPLR SAAGAAALAL TVDLPPASSE ARNSAFGFQG RLLHLSAGQR LGVHLHTEAR ARHAWQLTQG ATVLGLFRVT PEIPAGLPSP RSE | |
Greater than 80% as determined by SDS-PAGE. |
8744 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |