missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human 4-1BB/TNFRSF9/CD137 Protein

Product Code. 18427011 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427011 100 μg 100µg
18406821 500 μg 500µg
18439811 1 mg 1mg
18445981 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18427011 Supplier Novus Biologicals™ Supplier No. NBP234958100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

CD137 protein is fully biologically active when compared to standard. The biological activity is determined by its inhibitory effect of IL-8 production using human peripheral blood mononuclear cells. About 90 % of inibition was seen using a concentration of 1 μg for both 4-1BB Ligand and 4-1BB Receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07011, Q07011, Q07011
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 10 mM PB, pH 8.0, 150 mM NaCl.
Gene ID (Entrez) 3604
Molecular Weight (g/mol) 17.7 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of 4-1BB/TNFRSF9/CD137 as determined by LAL method.
Gene Alias 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Gene Symbol TNFRSF9
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells
Purity or Quality Grade >97% pure by SDS-PAGE and HPLC
Protein 4-1BB/TNFRSF9/CD137
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.