missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human 4-1BB Ligand/TNFSF9 Protein

Product Code. 18187280
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and PEG and with Trehalose.
Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and PEG with BSA as a carrier protein.
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18187280

Brand: R&D Systems™ 22954L025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Human (rh) 4-1BB/TNFRSF9/CD137 Fc Chimera (Catalog # 838-4B ) is Immobilized at 10 ng/mL (100 μL/well), the concentration of rh4-1BB Ligand that produces 50% optimal binding response is found to be approximately 0.5-2.5 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P41273.1
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and PEG and with Trehalose.
Gene ID (Entrez) 8744
Molecular Weight (g/mol) M.W. (theoretical): 20.4 kDa
Quantity 25 μg
Source E. coli-derived human 4-1 BB Ligand/TNFSF9 protein Arg71-Glu254, with an N-terminal Met and a 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, 2°C to 8°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFSF9
Product Type 4-1BB Ligand/TNFSF9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein 4-1 BB Ligand/TNFSF9
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.