missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
Fully biologically active when compared to standard. The ED50 as determined by stimulation of IL-8 production using human PBMC is less than 10 ng/mL, corresponding to a specific activity of >1.0 x 10^5 IU/mg.
Specifica
Specifica
| Accession Number | P41273, P41273, P41273 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 8744 |
| Molecular Weight (g/mol) | 19.4 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of 4-1BB Ligand/TNFSF9 as determined by LAL method. |
| Gene Alias | 4-1BB ligand, 4-1BBL, 4-1BB-L, 4-1BBL/TNFSF9, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?