missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L42, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L42, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6382-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_054769.1 | |
39S ribosomal protein L42, mitochondrial Protein | |
Research Use Only | |
MRPL42 | |
Recombinant Protein | |
E. coli | |
KSTYSPLPDD YNCNVELALT SDGRTIVCYH PSVDIPYEHT KPIPRPDPVH NNEETHDQVL KTRLEEKVEH LEEGPMIEQL SKMFFTTKHR WYPHGRYHRC RKNLNPPKDR | |
Greater than 90% as determined by SDS-PAGE. |
28977 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |