missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L28, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L28, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7579-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006419.2 | |
39S ribosomal protein L28, mitochondrial Protein | |
Research Use Only | |
MRPL28 | |
Recombinant Protein | |
E. coli | |
MPLHKYPVWL WKRLQLREGI CSRLPGYYLR SLEEERTPTP VHYRPHGAKF KINPKNGQRE RVEDVPIPIY FPPESQRGLW GGEGWILGQI YANNDKLSKR LKKVWKPQLF EREFYSEILD KKFTVTVTMR TLDLIDEAYG LDFYILKTPK EDLCSKFGMD LKRGMLLRLA RQDPQLHPED PERRAAIYDK YKEFAIPEEE AEWVGLTLEE AIEKQRLLEE KDPVPLFKIY VAELIQQLQQ QALSEPAVVQ KRASGQ | |
Greater than 80% as determined by SDS-PAGE. |
10573 | |
500 μg | |
Not determined. | |
Full Length of BC000990 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |