missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 39S ribosomal protein L11, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L11, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8279-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16073731

  • £384.00 / 100µg
Estimated Shipment: 16-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_057134.1
39S ribosomal protein L11, mitochondrial Protein
Research Use Only
MRPL11
Recombinant Protein
E. coli
MSKLGRAARG LRKPEVGGVI RAIVRAGLAM PGPPLGPVLG QRGVSINQFC KEFNERTKDI KEGIPLPTKI LVKPDRTFEI KIGQPTVSYF LKAAAGIEKG ARQTGKEVAG LVTLKHVYEI ARIKAQDEAF ALQDVPLSSV VRSIIGSARS LGIRVVKDLS SEELAAFQKE RAIFLAAQKE ADLAAQEEAA K
Greater than 90% as determined by SDS-PAGE.
65003
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers