missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 28S ribosomal protein S22, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 28S ribosomal protein S22, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6385-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_064576.1 | |
28S ribosomal protein S22, mitochondrial Protein | |
Research Use Only | |
MRPS22 | |
Recombinant Protein | |
E. coli | |
MAPLGTTVLL WSLLRSSPGV ERVCFRARIQ PWHGGLLQPL PCSFEMGLPR RRFSSEAAES GSPETKKPTF MDEEVQSILT KMTGLNLQKT FKPAIQELKP PTYKLMTQAQ LEEATRQAVE AAKVRLKMPP VLEERVPIND VLAEDKILEG TETTKYVFTD ISYSIPHRER FIVVREPSGT LRKASWEERD RMIQVYFPKE GRKILTPIIF KEENLRTMYS QDRHVDVLNL CFAQFEPDST EYIKVHHKTY EDIDKRGKYD LLRSTRYFGG MVWYFVNNKK IDGLLIDQIQ RDLIDDATNL VQLYHVLHPD GQSAQGAKDQ AAEGINLIKV FAKTEAQKGA YIELTLQTYQ EALSRHSAAS | |
Greater than 80% as determined by SDS-PAGE. |
56945 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |