missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human 14-3-3 tau/theta Protein

Product Code. 18078394 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18078394

Brand: Novus Biologicals™ NBC118393

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids1-245 of Human 14-3-3 tau/theta The Recombinant Human 14-3-3 tau/theta Protein is derived from E. coli. The Recombinant Human 14-3-3 tau/theta Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27348
For Use With (Application) Western Blot
Formulation PBS (pH 7.4), 10% glycerol
Gene ID (Entrez) 10971
Molecular Weight (g/mol) 28kDa
Name 14-3-3 tau/theta Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 14-3-3 tau/theta, 1-245aa, Human, Recombinant, E.coli (NP_006817).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.