missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HSV HCV Core 169aa Protein

A cDNA sequence encoding the HCV Core 169aa was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12141-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15969498

  • £1324.47 / 1mg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSV HCV Core 169aa Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HSV
His
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
Sterile filtered solution containing Phosphate saline buffer, 50mM arginine and 1M urea.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers