missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV Type-O gp41 13kDa Protein

A cDNA sequence encoding the HIV Type-O gp41 13kDa was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12239-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939798

  • £660.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV Type-O gp41 13kDa Protein
Research Use Only
Recombinant Protein
E. coli
Lyophilized from 1 mg/ml in 20mM Na-carbonate, pH 9.6.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
His
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers