missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV HTLV-1 p24 Protein

A cDNA sequence encoding the HTLV-1 p24 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12357-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15970159

  • £660.00 / 500µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV HTLV-1 p24 Protein
Research Use Only
Recombinant Protein
E. coli
HTLV-1 p24 protein solution containing PBS, pH 7.4.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Protein is >95% pure as determined by 12% SDS-PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers