missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 TAT Cys22 Protein
A cDNA sequence encoding the HIV-1 TAT Cys22 was constructed and used to recombinantly synthesize the protein.
£104.00 - £1165.00
Specifications
Name | HIV-1 TAT Cys22 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | Untagged |
Host Species | E. coli |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959878
|
enQuireBio™
QP12266-2UG |
2 μg |
£104.00
2µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15949878
|
enQuireBio™
QP12266-10UG |
10 μg |
£159.00
10µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15939878
|
enQuireBio™
QP12266-100UG |
100 μg |
£1165.00
100µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
HIV-1 TAT Cys22 Protein | |
Untagged | |
HIV |
Recombinant Protein | |
E. coli | |
Greater than 90.0% as determined by SDS-PAGE. |