missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 p24 Core Protein
A cDNA sequence encoding the HIV-1 p24 Core was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12256-EC-100ug
Additional Details : Weight : 0.01000kg
Specifications
HIV-1 p24 Core Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein solution contains 10mM Tris pH 8.0, 300mM NaCl and 0.01% Tween-20. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 90.0% as determined by HPLC analysis & SDS-PAGE. |