missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 p24 Core Protein

A cDNA sequence encoding the HIV-1 p24 Core was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12256-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929848

  • £99.15 / 2µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV-1 p24 Core Protein
Research Use Only
Recombinant Protein
E. coli
The protein solution contains 10mM Tris pH 8.0, 300mM NaCl and 0.01% Tween-20.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Greater than 90.0% as determined by HPLC analysis & SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers