missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5 Protein
A cDNA sequence encoding the HCV NS5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12185-500ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C HCV NS5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M Urea, 25mM Tris-HCl pH 8.0, 50% glycerol and 0.2% Triton-X. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
HCV-NS5 protein is >95% pure as determined by 10% PAGE (coomassie staining). |