missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-2c Protein

A cDNA sequence encoding the HCV NS3 Genotype-2c was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12168-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979578

  • £1320.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis C HCV NS3 Genotype-2c Protein
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
HCV NS3 Genotype-2c protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers