missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS3 Genotype-2c Protein

A cDNA sequence encoding the HCV NS3 Genotype-2c was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12168-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15969578

  • £170.83 / 100µg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis C HCV NS3 Genotype-2c Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
HCV NS3 Genotype-2c protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers