missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV Mosaic-B Protein

A cDNA sequence encoding the HCV Mosaic-B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12161-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979558

  • £660.00 / 500µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis C HCV Mosaic-B Protein
Research Use Only
Recombinant Protein
E. coli
HCV Mosaic-B protein solution containing 50mM arginine in PBS.
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers