missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-5 Protein
A cDNA sequence encoding the HCV Core Genotype-5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12154-500ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C HCV Core Genotype-5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, pH 8.0, 1.5M Urea, 0.2% Triton-X and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
Protein is >95% pure as determined by SDS-PAGE. |