missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV Core 24 Protein

A cDNA sequence encoding the HCV Core 24 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12143-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929508

  • £1320.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hepatitis C HCV Core 24 Protein
Research Use Only
Recombinant Protein
E. coli
20mM Tris-Hcl pH 8.0, 1.5M urea and 10mM b-mercaptoethanol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
HCV Core 24 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers