missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis B HBV X Protein

Product Code. 15949478
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
This item is not returnable. View return policy

Product Code. 15949478

Brand: enQuireBio™ QP121341mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the HBV X was constructed and used to recombinantly synthesize the protein.

Specifications

Name Hepatitis B HBV X Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol X
Product Type Recombinant Protein
Cross Reactivity Hepatitis B
Species E. coli
Protein Tag Untagged
Sequence MAARVCCQLD PARDVLCLRP VGAESRGRPV SGPFGTLPSP SSSAVPADHG AHLSLRGLPV CAFSSAGPCA LRFTSARRME TTVNAHQVLP KVLHKRTLGL SAMSTTDLEA YFKDCLFKDW EELGEEIRLK VFVLGGCRHK LVCSPAPCNF FTSA
Buffer Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 30mM acetate buffer pH4.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.