missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg ayw Protein
A cDNA sequence encoding the HBsAg ayw was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | Hepatitis B HBsAg ayw Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15939458
|
enQuireBio™
QP12127-HIS-EC-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959458
|
enQuireBio™
QP12127-HIS-EC-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949458
|
enQuireBio™
QP12127-HIS-EC-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis B HBsAg ayw Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HBsAg solution (1 mg/ml) containing 7.5mM phosphate buffer pH 7.2, 75mM NaCl and 50% glycerol. |