missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis A HAV P3C Protein

Product Code. 15969398
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 15969398

Brand: enQuireBio™ QP12110100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the HAV P3C was constructed and used to recombinantly synthesize the protein.

Specifications

Name Hepatitis A HAV P3C Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Hepatitis A
Species E. coli
Protein Tag Untagged
Buffer 10mM CBB, pH9.6, 0.1% SDS and 50% glycerol.
Purity or Quality Grade HAV P3C protein is >90% pure as determined by 10% PAGE(coomassie staining).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.